Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016892.3 | NP_058588.1 | 7 | 734 | ||
GenBank mRNA | AF121906.1 | - | 7 | 647 | ||
AF173379.1 | - | 7 | 707 | |||
AK010264.1 | - | 4 | 343 | |||
AK050072.1 | - | 7 | 683 | |||
BC026938.1 | - | 7 | 701 | 4024140 | ||
CF929049.1 | - | 1 | 60 |