Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007657.3 | NP_031683.1 | 7 | 679 | ||
GenBank mRNA | AK002251.1 | - | 7 | 685 | ||
AK012793.1 | - | 7 | 685 | |||
AK151443.1 | - | 7 | 645 | |||
AK151619.1 | - | 7 | 632 | |||
AK170310.1 | - | 7 | 632 | |||
BC070474.1 | - | 7 | 679 | 30634238 | ||
L08115.1 | - | 7 | 617 |