Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243769.1 | NP_001230698.1 | 5 | 499 | ||
NM_009861.3 | NP_033991.1 | 5 | 499 | |||
GenBank mRNA | AK003098.1 | - | 5 | 420 | ||
AK051543.1 | - | 5 | 420 | |||
AK075567.1 | - | 5 | 418 | |||
AK144216.1 | - | 5 | 422 | |||
AK145659.1 | - | 4 | 263 | |||
AK150563.1 | - | 5 | 432 | |||
AK151087.1 | - | 5 | 444 | |||
AK151726.1 | - | 5 | 421 | |||
AK153564.1 | - | 5 | 443 | |||
AK154870.1 | - | 5 | 418 | |||
AK159470.1 | - | 5 | 461 | |||
AK166281.1 | - | 5 | 418 | |||
AK167195.1 | - | 5 | 407 | |||
AK167400.1 | - | 5 | 412 | |||
AK167609.1 | - | 5 | 417 | |||
AK168013.1 | - | 4 | 412 | |||
AK168089.1 | - | 4 | 412 | |||
AK168276.1 | - | 5 | 412 | |||
AK168503.1 | - | 5 | 433 | |||
AK168758.1 | - | 5 | 452 | |||
AK168820.1 | - | 5 | 425 | |||
AK169122.1 | - | 5 | 433 | |||
AK169232.1 | - | 5 | 412 | |||
AK169805.1 | - | 5 | 437 | |||
AK181967.1 | - | 2 | 130 | |||
AK213611.1 | - | 4 | 273 | |||
AK218916.1 | - | 2 | 130 | |||
BC064792.1 | - | 5 | 405 | 30105331 | ||
CD353254.1 | - | 2 | 83 | |||
CD551112.1 | - | 5 | 486 | |||
L11318.1 | - | 4 | 335 | |||
L78075.1 | - | 4 | 339 | |||
U37720.1 | - | 5 | 407 |