Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278041.1 | NP_001264970.1 | 16 | 2181 | ||
NM_009893.2 | NP_034023.1 | 16 | 2115 | |||
GenBank mRNA | AF069501.1 | - | 16 | 2115 | ||
AF096276.1 | - | 16 | 2094 | |||
AK077460.1 | - | 17 | 2276 | |||
AK149445.1 | - | 16 | 2090 | |||
AK220310.1 | - | 2 | 2276 | |||
BC145017.1 | - | 15 | 2323 | |||
BC145018.1 | - | 17 | 2243 | |||
BC145019.1 | - | 17 | 2280 | |||
BC145696.1 | - | 16 | 2179 | 40130999 |