Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007710.2 | NP_031736.1 | 2 | 236 | ||
GenBank mRNA | AK009950.1 | - | 2 | 237 | ||
AK137460.1 | - | 238 | ||||
BC132424.1 | - | 2 | 234 | 40130701 | ||
BC132426.1 | - | 2 | 234 | 40130703 | ||
X03233.1 | - | 2 | 236 |