Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016675.4 | NP_057884.1 | 2 | 798 | ||
GenBank mRNA | AF072128.1 | - | 1 | 460 | ||
AK004990.1 | - | 1 | 565 | |||
BC015252.1 | - | 1 | 538 | 4223446 | ||
BC085494.1 | - | 2 | 778 | 5686202 |