Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009950.2 | NP_034080.1 | 3 | 467 | ||
GenBank mRNA | AF017992.1 | - | 2 | 348 | ||
AJ224738.1 | - | 3 | 466 | |||
AJ224739.1 | - | 2 | 203 | |||
AJ224740.1 | - | 3 | 452 | |||
AK136561.1 | - | 3 | 372 | |||
AK163112.1 | - | 3 | 479 | |||
BC005608.1 | - | 2 | 430 | 3498522 |