Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007867.4 | NP_031893.3 | 2, 3 | 788 | ||
GenBank mRNA | AK145123.1 | - | 2, 3 | 1251 | ||
BC106967.2 | - | 2, 3 | 468 | 40040019 | ||
U73329.1 | - | 2, 3 | 468 |