Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080388.2 | NP_001073857.1 | 17 | 2676 | ||
NM_001080389.2 | NP_001073858.1 | 18 | 2703 | |||
NM_001080390.2 | NP_001073859.1 | 18 | 2555 | |||
NM_007928.3 | NP_031954.2 | 18 | 2838 | |||
GenBank mRNA | AK045329.1 | - | 18 | 2412 | ||
AK154864.1 | - | 18 | 2555 | |||
AK172444.1 | - | 18 | 2663 | |||
AK172664.1 | - | 18 | 2481 | |||
AK220191.1 | - | 19 | 2404 | |||
BC058556.1 | - | 17 | 2613 | 5289799 | ||
DV650107.1 | - | 5 | 645 | |||
X70764.1 | - | 18 | 2286 |