Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021371.2 | NP_067346.1 | 4 | 1001 | ||
NM_181045.1 | NP_851388.1 | 5 | 701 | |||
GenBank mRNA | AB041539.1 | - | 4 | 992 | ||
AF282251.1 | - | 5 | 683 | |||
AK035865.1 | - | 5 | 670 | |||
AK081011.1 | - | 4 | 435 | |||
AK158641.1 | - | 4 | 363 | |||
AK158892.1 | - | 4 | 973 | |||
AK163648.1 | - | 4 | 464 | |||
BC057129.1 | - | 4 | 980 | 6811387 | ||
BC058963.1 | - | 4 | 980 | 6811387 |