Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010185.4 | NP_034315.1 | 5 | 333 | ||
GenBank mRNA | AK155600.1 | - | 5 | 290 | ||
BC034163.1 | - | 273 | 5065647 | |||
BE634435.1 | - | 1 | 320 | |||
BY173505.1 | - | 5 | 333 | |||
CK791068.1 | - | 5 | 283 | |||
J05020.1 | - | 5 | 271 |