Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008081.3 | NP_032107.1 | 9 | 1041 | ||
GenBank mRNA | BC071231.1 | - | 9 | 1009 | ||
BC139165.1 | - | 9 | 1031 | 8862187 | ||
BC139166.1 | - | 9 | 1031 | 8862188 | ||
L30104.1 | - | 9 | 1041 |
Gene Symbol: 4833417C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133925.1 | - | 793 |