Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008092.3 | NP_032118.2 | 5 | 1571 | ||
GenBank mRNA | AB075549.1 | - | 5 | 1589 | ||
AF179424.1 | - | 4 | 992 | |||
AK134639.1 | - | 4 | 595 | |||
BC137824.1 | - | 4 | 1324 | 8860843 | ||
M98339.1 | - | 5 | 1539 | |||
U85046.1 | - | 5 | 1542 |