Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010316.3 | NP_034446.1 | 2 | 313 | ||
GenBank mRNA | AK004459.1 | - | 2 | 278 | ||
AK032646.1 | - | 2 | 261 | |||
BC029680.1 | - | 2 | 197 | 5365311 | ||
U38497.1 | - | 1 | 20 |