Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010359.2 | NP_034489.1 | 5 | 359 | ||
GenBank mRNA | BC044927.1 | - | 5 | 343 | 5051815 | |
J03953.1 | - | 4 | 299 |