Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-Ab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207105.3 | NP_996988.2 | 3 | 604 | ||
GenBank mRNA | AF065912.1 | - | 3 | 507 | ||
AF065913.1 | - | 3 | 534 | |||
AF119251.1 | - | 3 | 513 | |||
AF119252.1 | - | 3 | 513 | |||
AK077436.1 | - | 3 | 592 | |||
AK155573.1 | - | 3 | 576 | |||
AK156899.1 | - | 3 | 576 | |||
AK170370.1 | - | 3 | 586 | |||
AK171005.1 | - | 3 | 576 | |||
AK171211.1 | - | 3 | 580 | |||
AK171281.1 | - | 3 | 576 | |||
AK171290.1 | - | 3 | 576 | |||
AK171298.1 | - | 3 | 576 | |||
AK172409.1 | - | 3 | 577 | |||
AY303784.1 | - | 2 | 437 | |||
AY303785.1 | - | 2 | 428 | |||
AY452202.1 | - | 3 | 513 | |||
BC008168.1 | - | 3 | 596 | 3257494 | ||
BC010322.1 | - | 3 | 570 | 2651058 | ||
BC057998.1 | - | 3 | 562 | 5340289 | ||
CK330363.1 | - | 4 | 578 | |||
M13537.1 | - | 3 | 519 | |||
M13539.1 | - | 3 | 513 | |||
M13541.1 | - | 3 | 481 | |||
M15848.1 | - | 3 | 549 | |||
M66213.1 | - | 3 | 513 |