Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010442.2 | NP_034572.1 | 2, 3 | 279 | ||
GenBank mRNA | AK150934.1 | - | 2, 3 | 276 | ||
AK151005.1 | - | 2, 3 | 276 | |||
AK151012.1 | - | 2, 3 | 267 | |||
AK151627.1 | - | 2, 3 | 254 | |||
AK153420.1 | - | 2, 3 | 276 | |||
AK153516.1 | - | 2, 3 | 276 | |||
AK153566.1 | - | 2, 3 | 276 | |||
AK154598.1 | - | 2, 3 | 277 | |||
AK159521.1 | - | 2, 3 | 276 | |||
AK159870.1 | - | 2, 3 | 280 | |||
AK159959.1 | - | 2, 3 | 280 | |||
AK169227.1 | - | 2, 3 | 276 | |||
BC010757.1 | - | 2, 3 | 248 | 4235374 | ||
BY319630.1 | - | 2, 3 | 281 | |||
M33203.1 | - | 2, 3 | 221 | |||
X13356.1 | - | 2, 3 | 221 |