Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifngr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008338.3 | NP_032364.1 | 2 | 240 | ||
GenBank mRNA | AK086929.1 | - | 2 | 254 | ||
AK137165.1 | - | 2 | 263 | |||
AK143214.1 | - | 2 | 255 | |||
AK146995.1 | - | 258 | ||||
AK165193.1 | - | 2 | 256 | |||
AK169907.1 | - | 2 | 260 | |||
BC055745.1 | - | 2 | 240 | 6403454 | ||
S69336.1 | - | 2 | 264 |