Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008344.3 | NP_032370.2 | 4 | 1033 | ||
GenBank mRNA | AK157982.1 | - | 4 | 717 | ||
AW123368.1 | - | 1 | 269 | |||
BC012723.1 | - | 4 | 740 | 4224052 | ||
CT010184.1 | - | 4 | 671 | |||
X81584.1 | - | 4 | 863 |