Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008344.3 | NP_032370.2 | 2 | 807 | ||
GenBank mRNA | AK157982.1 | - | 2 | 490 | ||
BC012723.1 | - | 2 | 514 | 4224052 | ||
CT010184.1 | - | 2 | 445 | |||
X81584.1 | - | 2 | 637 |