Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il18bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010531.1 | NP_034661.1 | 6 | 614 | ||
GenBank mRNA | AB019505.1 | - | 4 | 463 | ||
AF110802.1 | - | 4 | 1841 | |||
AF110803.1 | - | 5 | 541 | |||
AF122907.1 | - | 6 | 614 | |||
AK003370.1 | - | 6 | 630 | |||
AK008452.1 | - | 5 | 545 | |||
AK009721.1 | - | 5 | 541 | |||
AK009877.1 | - | 2 | 181 | |||
BC018332.1 | - | 5 | 524 | 4007411 | ||
CT010164.1 | - | 4 | 463 | |||
CT010334.1 | - | 4 | 463 |