Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010599.4 | NP_034729.3 | 8 | 994 | ||
GenBank mRNA | AK029750.1 | - | 7 | 583 | ||
AK031806.1 | - | 8 | 975 | |||
AK044102.1 | - | 8 | 949 | |||
AK046194.1 | - | 8 | 984 | |||
AK049316.1 | - | 8 | 995 | |||
AK083134.1 | - | 8 | 996 | |||
BC017518.1 | - | 8 | 920 | 4507029 | ||
U65593.1 | - | 1 | 105 |