Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010663.2 | NP_034793.1 | 6, 7 | 1212 | ||
GenBank mRNA | AB013608.1 | - | 6, 7 | 1213 | ||
BC032161.1 | - | 6, 7 | 1259 | |||
BC132454.1 | - | 6, 7 | 1247 | 40130731 | ||
BC132456.1 | - | 6, 7 | 1247 | 40130733 | ||
M13805.1 | - | 3, 4 | 504 |