Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008496.4 | NP_032522.2 | 2 | 155 | ||
GenBank mRNA | AF331640.1 | - | 1 | 60 | ||
AK003153.1 | - | 2 | 89 | |||
AK008773.1 | - | 2 | 89 | |||
AK009151.1 | - | 2 | 98 | |||
BC116838.1 | - | 2 | 116 | 40126157 | ||
BC116840.1 | - | 2 | 116 | 40126159 | ||
BG803457.1 | - | 2 | 312 |