Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mesp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008589.2 | NP_032615.2 | 2 | 1265 | ||
GenBank mRNA | AK146740.1 | - | 2 | 745 | ||
BC145652.1 | - | 2 | 1095 | 40130955 | ||
BC145654.1 | - | 2 | 1095 | 40130957 | ||
U71125.1 | - | 2 | 1270 |