Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008612.2 | NP_032638.2 | 2 | 280 | ||
GenBank mRNA | AK008134.1 | - | 2 | 245 | ||
AK008249.1 | - | 2 | 283 | |||
AK011833.1 | - | 2 | 241 | |||
AK029174.1 | - | 2 | 192 | |||
AK048499.1 | - | 2 | 286 | |||
AK155859.1 | - | 2 | 281 | |||
AK157721.1 | - | 2 | 267 | |||
BC089023.1 | - | 2 | 269 | 30702121 | ||
BC115630.2 | - | 2 | 272 | 40102880 | ||
BC115631.1 | - | 2 | 273 | |||
U35249.1 | - | 2 | 311 |