Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK010218.1 | - | 2 | 326 | ||
AK038631.1 | - | 2 | 374 | |||
AK156731.1 | - | 2 | 300 | |||
AK202557.1 | - | 1 | 61 | |||
AY176058.1 | - | 2 | 333 | |||
AY176059.1 | - | 2 | 364 | |||
BC021938.1 | - | 2 | 331 | 5136809 | ||
U19799.1 | - | 2 | 345 |