Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010914.2 | NP_035044.1 | 6, 7 | 715 | ||
GenBank mRNA | AK010762.1 | - | 6, 7 | 695 | ||
AK011792.1 | - | 6, 7 | 634 | |||
AK079209.1 | - | 6, 7 | 670 | |||
AK146168.1 | - | 6, 7 | 717 | |||
BC010719.1 | - | 6, 7 | 656 | 2811674 | ||
X55316.1 | - | 6, 7 | 711 |