Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010924.2 | NP_035054.1 | 3 | 524 | ||
GenBank mRNA | AK006371.1 | - | 2 | 271 | ||
BC028757.1 | - | 3 | 440 | 1067421 | ||
BX633297.1 | - | 1 | 430 | |||
U86105.1 | - | 3 | 571 | |||
U86106.1 | - | 3 | 571 | |||
U86107.1 | - | 3 | 571 | |||
U86108.1 | - | 3 | 571 |