Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163662.1 | NP_001157134.1 | 6 | 643 | ||
NM_008749.2 | NP_032775.1 | 6 | 643 | |||
GenBank mRNA | AK002630.1 | - | 6 | 624 | ||
AK004886.1 | - | 6 | 614 | |||
AK007976.1 | - | 6 | 617 | |||
AK045087.1 | - | 6 | 643 | |||
AK140710.1 | - | 6 | 621 | |||
AK160981.1 | - | 6 | 630 | |||
AK164088.1 | - | 6 | 625 | |||
AK170717.1 | - | 6 | 638 | |||
AK182386.1 | - | 2 | 108 | |||
BC072554.1 | - | 6 | 621 | 5684979 | ||
M96823.1 | - | 5 | 565 |