Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163662.1 | NP_001157134.1 | 7, 8 | 843 | ||
NM_008749.2 | NP_032775.1 | 7, 8 | 843 | |||
GenBank mRNA | AK002630.1 | - | 7, 8 | 824 | ||
AK004886.1 | - | 7, 8 | 814 | |||
AK007976.1 | - | 7, 8 | 817 | |||
AK045087.1 | - | 7, 8 | 843 | |||
AK140710.1 | - | 7, 8 | 821 | |||
AK160981.1 | - | 7, 8 | 830 | |||
AK164088.1 | - | 7, 8 | 825 | |||
AK170717.1 | - | 7, 8 | 838 | |||
AK194015.1 | - | 2, 3 | 111 | |||
AK209002.1 | - | 1, 2 | 49 | |||
BC072554.1 | - | 7, 8 | 821 | 5684979 | ||
M96823.1 | - | 6, 7 | 765 |