Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203507.3 | NP_987103.1 | 1, 2 | 302 | ||
GenBank mRNA | AK004597.1 | - | 1, 2 | 256 | ||
AK005797.1 | - | 1, 2 | 287 | |||
AK016505.1 | - | 1, 2 | 278 | |||
BB610203.1 | - | 1, 2 | 303 | |||
BC016198.1 | - | 1, 2 | 238 | 4503404 |