Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203507.3 | NP_987103.1 | 2 | 343 | ||
GenBank mRNA | AK004597.1 | - | 2 | 297 | ||
AK005797.1 | - | 2 | 328 | |||
AK016505.1 | - | 2 | 319 | |||
BB610203.1 | - | 2 | 344 | |||
BC016198.1 | - | 2 | 279 | 4503404 |