Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203507.3 | NP_987103.1 | 4 | 508 | ||
GenBank mRNA | AK004597.1 | - | 4 | 462 | ||
AK005797.1 | - | 4 | 493 | |||
AK016505.1 | - | 4 | 482 | |||
AK178835.1 | - | 3 | 161 | |||
AK185756.1 | - | 3 | 161 | |||
BC016198.1 | - | 4 | 444 | 4503404 |