Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011201.3 | NP_035331.3 | 4, 5 | 510 | ||
GenBank mRNA | AK149979.1 | - | 4, 5 | 466 | ||
AK150322.1 | - | 4, 5 | 466 | |||
AK151371.1 | - | 4, 5 | 423 | |||
AK153088.1 | - | 4, 5 | 449 | |||
AK156281.1 | - | 4, 5 | 471 | |||
AK157431.1 | - | 4, 5 | 511 | |||
AK165949.1 | - | 5, 6 | 590 | |||
AK166020.1 | - | 4, 5 | 466 | |||
AK168255.1 | - | 4, 5 | 448 | |||
AK168772.1 | - | 4, 5 | 465 | |||
AK170620.1 | - | 4, 5 | 466 | |||
AK171380.1 | - | 4, 5 | 466 | |||
AK172206.1 | - | 5, 6 | 583 | |||
BC010191.1 | - | 4, 5 | 428 | 3156854 | ||
M97590.1 | - | 4, 5 | 426 | |||
Z23057.1 | - | 1, 2 | 40 |