Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011201.3 | NP_035331.3 | 3, 4 | 416 | ||
GenBank mRNA | AK149979.1 | - | 3, 4 | 372 | ||
AK150322.1 | - | 3, 4 | 372 | |||
AK151371.1 | - | 3, 4 | 329 | |||
AK153088.1 | - | 3, 4 | 355 | |||
AK156281.1 | - | 3, 4 | 377 | |||
AK157431.1 | - | 3, 4 | 417 | |||
AK165567.1 | - | 3, 4 | 381 | |||
AK165949.1 | - | 4, 5 | 496 | |||
AK166020.1 | - | 3, 4 | 372 | |||
AK168255.1 | - | 3, 4 | 354 | |||
AK168772.1 | - | 3, 4 | 371 | |||
AK170620.1 | - | 3, 4 | 372 | |||
AK171380.1 | - | 3, 4 | 372 | |||
AK172206.1 | - | 4, 5 | 489 | |||
AK185938.1 | - | 2, 3 | 123 | |||
BC010191.1 | - | 3, 4 | 334 | 3156854 | ||
M97590.1 | - | 3, 4 | 332 |