Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011201.3 | NP_035331.3 | 7 | 897 | ||
GenBank mRNA | AK149979.1 | - | 7 | 853 | ||
AK150322.1 | - | 7 | 853 | |||
AK151371.1 | - | 7 | 810 | |||
AK153088.1 | - | 7 | 835 | |||
AK156281.1 | - | 7 | 859 | |||
AK157431.1 | - | 7 | 898 | |||
AK165567.1 | - | 7 | 862 | |||
AK165949.1 | - | 8 | 977 | |||
AK166020.1 | - | 7 | 853 | |||
AK168772.1 | - | 7 | 852 | |||
AK170620.1 | - | 7 | 853 | |||
AK171380.1 | - | 7 | 853 | |||
AK172206.1 | - | 8 | 970 | |||
AK208930.1 | - | 2 | 80 | |||
BC005729.1 | - | 2 | 250 | |||
BC010191.1 | - | 7 | 815 | 3156854 | ||
M97590.1 | - | 7 | 813 |