Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Relb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290457.1 | NP_001277386.1 | 6, 7 | 996 | ||
NM_009046.2 | NP_033072.2 | 7, 8 | 1005 | |||
GenBank mRNA | AK146932.1 | - | 7, 8 | 1006 | ||
AK156767.1 | - | 7, 8 | 961 | |||
BC019765.1 | - | 7, 8 | 1005 | 3584200 | ||
BC034523.1 | - | 6, 7 | 809 | 4485540 | ||
BC117792.2 | - | 5, 6 | 1002 | |||
BC117793.1 | - | 7, 8 | 941 | 40092162 | ||
DQ020178.1 | - | 6, 7 | 657 | |||
DQ020179.1 | - | 6, 7 | 1773 | |||
DQ020180.1 | - | 8, 9 | 1813 | |||
DQ020181.1 | - | 7, 8 | 1839 | |||
M83380.1 | - | 7, 8 | 842 |