Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009113.4 | NP_033139.2 | 4 | 526 | ||
GenBank mRNA | AK002755.1 | - | 3 | 325 | ||
AK008895.1 | - | 3 | 325 | |||
BC005687.1 | - | 4 | 526 | 3155915 | ||
CK334302.1 | - | 3 | 493 | |||
CX219167.1 | - | 3 | 249 | |||
X99921.1 | - | 3 | 253 |