Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC061126.1 | - | 3 | 291 | 30276350 | |
S50588.1 | - | 3 | 241 | |||
Z12297.1 | - | 3 | 241 |