Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013654.3 | NP_038682.1 | 2 | 206 | ||
GenBank mRNA | AF128193.1 | - | 2 | 171 | ||
AF128194.1 | - | 2 | 171 | |||
AK078827.1 | - | 2 | 207 | |||
BC061126.1 | - | 2 | 204 | 30276350 | ||
BY163435.1 | - | 2 | 208 | |||
L04694.1 | - | 2 | 168 | |||
S50588.1 | - | 2 | 154 | |||
S71251.1 | - | 2 | 186 | |||
Z12297.1 | - | 2 | 154 |