Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178661.4 | NP_848776.2 | 4 | 861 | ||
GenBank mRNA | AK035695.1 | - | 4 | 851 | ||
AK036712.1 | - | 4 | 853 | |||
AK041050.1 | - | 4 | 834 | |||
AK041669.1 | - | 4 | 861 | |||
AK042418.1 | - | 4 | 856 | |||
BC138470.1 | - | 4 | 707 | 8861490 | ||
BC138471.1 | - | 4 | 707 | 8861491 |