Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153056.2 | NP_694696.2 | 7 | 633 | ||
GenBank mRNA | AB221687.1 | - | 3 | 153 | ||
AK135484.1 | - | 6 | 703 | |||
AY251540.1 | - | 7 | 609 | |||
BC026403.1 | - | 7 | 598 | 4209498 | ||
BC026637.1 | - | 6 | 674 | |||
BC026650.1 | - | 7 | 601 | 4987746 | ||
BC040759.1 | - | 7 | 598 | 4209498 | ||
BC080304.1 | - | 6 | 937 |