Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009301.2 | NP_033327.1 | 2 | 375 | ||
GenBank mRNA | AK007282.1 | - | 2 | 376 | ||
BC114349.1 | - | 2 | 379 | 40053311 | ||
S60687.1 | - | 1 | 265 | |||
X57139.1 | - | 2 | 403 | |||
X63161.1 | - | 2 | 355 |