Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105196.1 | NP_001098666.1 | 9 | 1742 | ||
NM_001105197.1 | NP_001098667.1 | 10 | 1533 | |||
NM_001271489.1 | NP_001258418.1 | 10 | 1828 | |||
NM_001271490.1 | NP_001258419.1 | 10 | 2000 | |||
NM_001271491.1 | NP_001258420.1 | 9 | 1462 | |||
NM_172472.3 | NP_766060.2 | 10 | 1847 | |||
GenBank mRNA | AK089762.1 | - | 8 | 1504 | ||
AK139756.1 | - | 9 | 1739 | |||
AK144657.1 | - | 10 | 1812 | |||
AK144751.1 | - | 10 | 1816 | |||
AK145029.1 | - | 10 | 1969 | |||
AK145832.1 | - | 10 | 1812 | |||
AK147405.1 | - | 7 | 4906 | |||
AK150372.1 | - | 10 | 1599 | |||
AK151447.1 | - | 10 | 1518 | |||
AK152221.1 | - | 10 | 1518 | |||
AK153051.1 | - | 10 | 1599 | |||
AK155883.1 | - | 9 | 1464 | |||
AK159162.1 | - | 10 | 1727 | |||
BC056358.1 | - | 10 | 1835 | 6813486 | ||
BC063047.1 | - | 10 | 1835 | 6813486 | ||
S76673.1 | - | 8 | 1185 |