Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105196.1 | NP_001098666.1 | 7 | 1268 | ||
NM_001105197.1 | NP_001098667.1 | 8 | 1059 | |||
NM_001271489.1 | NP_001258418.1 | 8 | 1354 | |||
NM_001271490.1 | NP_001258419.1 | 8 | 1526 | |||
NM_001271491.1 | NP_001258420.1 | 7 | 988 | |||
NM_172472.3 | NP_766060.2 | 8 | 1373 | |||
GenBank mRNA | AK089762.1 | - | 6 | 1030 | ||
AK139756.1 | - | 7 | 1265 | |||
AK144657.1 | - | 8 | 1338 | |||
AK144751.1 | - | 8 | 1341 | |||
AK145029.1 | - | 8 | 1494 | |||
AK145832.1 | - | 8 | 1338 | |||
AK147405.1 | - | 6 | 3054 | |||
AK150372.1 | - | 8 | 1125 | |||
AK151447.1 | - | 8 | 1044 | |||
AK152221.1 | - | 8 | 1044 | |||
AK153051.1 | - | 8 | 1125 | |||
AK154690.1 | - | 7 | 1924 | |||
AK155883.1 | - | 7 | 990 | |||
AK159162.1 | - | 8 | 1254 | |||
BC056358.1 | - | 8 | 1361 | 6813486 | ||
BC063047.1 | - | 8 | 1361 | 6813486 | ||
S76673.1 | - | 6 | 711 |