Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyw3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168358.1 | NP_001161830.1 | 5 | 569 | ||
NM_172474.5 | NP_766062.1 | 6 | 703 | |||
GenBank mRNA | AK030344.1 | - | 6 | 701 | ||
AK034820.1 | - | 6 | 688 | |||
BC086686.1 | - | 6 | 677 | 30680486 | ||
BC087879.1 | - | 6 | 667 | 6533208 | ||
BC141040.1 | - | 6 | 682 | 9055652 |