Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145964.2 | NP_666076.2 | 10 | 1046 | ||
GenBank mRNA | BB611022.1 | - | 10 | 734 | ||
BC002199.1 | - | 1 | 256 |