Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289468.1 | NP_001276397.1 | 2 | 289 | ||
NM_009337.3 | NP_033363.1 | 2 | 289 | |||
GenBank mRNA | AF031956.1 | - | 2 | 265 | ||
AK149361.1 | - | 2 | 271 | |||
AK163387.1 | - | 2 | 272 | |||
BC052336.1 | - | 2 | 270 | 30053730 | ||
CA558861.1 | - | 2 | 258 | |||
CJ087554.1 | - | 2 | 272 | |||
DV648028.1 | - | 2 | 248 | |||
DV653827.1 | - | 2 | 289 | |||
DV655843.1 | - | 2 | 282 | |||
Y15376.1 | - | 2 | 265 |