Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009337.3 | NP_033363.1 | 3 | 398 | ||
GenBank mRNA | AF031956.1 | - | 3 | 374 | ||
AK163387.1 | - | 3 | 381 | |||
BC052336.1 | - | 3 | 379 | 30053730 | ||
CA558861.1 | - | 3 | 318 | |||
CJ087217.1 | - | 2 | 209 | |||
DV648028.1 | - | 3 | 605 | |||
DV649899.1 | - | 2 | 198 | |||
DV653827.1 | - | 3 | 398 | |||
DV655843.1 | - | 3 | 342 | |||
DV663230.1 | - | 2 | 100 | |||
Y15376.1 | - | 3 | 374 |